Learn More
Indofine Chemical Isorhamnetin With , 98%, 10 Mg
SDP
Supplier:  Indofine Chemical 021120S 10 MG
Encompass_Preferred
ISORHAMNETIN with HPLC 10 MG, Flavone, 3,5,7,4'-Tetrahydroxy-3'-methoxyflavone, C16H12O7, MW: 316.27, 98+%, 480-19-3, MFCD00017310
Catalog No. 50-908-546
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.