Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isosorbide mononitrate, 98+%

Catalog No. AAJ6367409
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6367409 10 g
AAJ6367406 5 g
AAJ6367414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6367409 Supplier Thermo Scientific Chemicals Supplier No. J6367409
Add to Cart

A coronary vasodilator that does not require hepatic biotransformation | CAS: 16051-77-7 | C6H9NO6 | 191.139 g/mol

A coronary vasodilator that does not require hepatic biotransformation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16051-77-7
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.139
MDL Number MFCD00143462
InChI Key YWXYYJSYQOXTPL-SLPGGIOYSA-N
Synonym isosorbide mononitrate, isosorbide 5-mononitrate, monosorbitrate, imdur, elantan, monoket, mononit, corangin, ismn, isosorbide 5-nitrate
PubChem CID 27661
ChEBI CHEBI:6062
IUPAC Name [(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
SMILES C1C(C2C(O1)C(CO2)O[N+](=O)[O-])O

Specifications

Melting Point 92°C
Color White
Quantity 10 g
UN Number UN3251
Merck Index 14,5225
Formula Weight 191.14
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Isosorbide mononitrate
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: ISOSORBIDE-5-MONONITRATE
EINECSNumber 240-197-2
RTECSNumber LZ4386500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.