Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators

Catalog No. FBG30CSGA Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
27 cu. ft.
49 cu. ft.
72 cu. ft.
Shelves:
12 Full, 2 Half
4 Full, 3 Half
8 Full, 2 Half
Color:
Gray
Interior:
Stainless Steel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Shelves Color Interior
FBG30CSGA 27 cu. ft. 4 Full, 3 Half Gray Stainless Steel
FBG49CSGA 49 cu. ft. 8 Full, 2 Half Gray Stainless Steel
FBG72CSGA 72 cu. ft. 12 Full, 2 Half Gray Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. FBG30CSGA
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators are designed with your needs in mind.

  • Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
  • Whisper-quiet operation
  • ENERGY STAR certified
  • Temperature, door ajar, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
  • Internal plug/outlet
  • Special safety sensor turns the internal plug/outlet off if a hydrocarbon (coolant) leak is detected
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 27 cu. ft.
Capacity (Metric) 764 L
Temperature Range 2°C to 10°C
Temperature (Setpoint) 5°C
Refrigerant R290
Door Style Glass
No. of Doors 1
Shelves 4 Full, 3 Half
Capacity (English) Shelf 100 lb./full shelf; 50 lb./half shelf
Capacity (Metric) Shelf 45 kg/full shelf, 22.5 kg/half shelf
Interior Stainless Steel
Certifications/Compliance cULus, ENERGYSTAR
Color Gray
Control Type Microprocessor
Ports 1 Rear access port and 2 side access ports
Warranty 2 years parts and labor, plus 3 year compressor warranty
Model CSGA
Access Security Key Lock
Dimensions (D x W x H) Interior 28.5 x 27 x 61.75 in. (724 x 686 x 1569 mm)
Dimensions (L x W x H) Exterior 32 x 30.75 x 81 in.(813 x 781 x 813 mm)
Amperage 3 A
Plug Type NEMA 5-15 P
Electrical Requirements 115 V 60 Hz
Shipping Weight (English) 386 lb.
Shipping Weight (Metric) 175 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.