Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators

Catalog No. FBG25RPGA Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
12 cu. ft.
23 cu. ft.
49 cu. ft.
72 cu. ft.
Shelves:
12 Full
4 Full
8 Full
Color:
Gray
White
Interior:
Painted White Steel
Stainless Steel
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Shelves Color Interior
FBG25RPGA 23 cu. ft. 4 Full White Painted White Steel
FBV72RPGA 72 cu. ft. 12 Full White Painted White Steel
FBV72RSGA 72 cu. ft. 12 Full Gray Stainless Steel
FBG12RPGA 12 cu. ft. 4 Full White Painted White Steel
FBG49RPGA 49 cu. ft. 8 Full White Painted White Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. FBG25RPGA
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.

  • Whisper-quiet operation
  • ENERGY STAR certified
  • Temperature, door ajar, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 23 cu. ft.
Capacity (Metric) 651 L
Temperature Range 2°C to 10°C
Temperature (Setpoint) 5°C
Refrigerant R290
Door Style Glass
Shelves 4 Full
Capacity (English) Shelf 100 lb.
Capacity (Metric) Shelf 45 kg
Interior Painted White Steel
Certifications/Compliance cULus, ENERGYSTAR
Color White
Control Type Microprocessor
Ports 1 Rear access port
Warranty 13 months parts and labor
Model RPGA
Access Security Key Lock
Dimensions (D x W x H) Interior 28.5 x 24 x 61.75 in. (724 x 610 x 1569 mm)
Dimensions (L x W x H) Exterior 32 x 27.75 x 81 in.(813 x 705 x 2057 mm)
Amperage 5.8 A
Plug Type NEMA 5-15 P
Electrical Requirements 115 V 60 Hz
Shipping Weight (English) 392 lb.
Shipping Weight (Metric) 178 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.