Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isothiazole 98.0+%, TCI America™
SDP

Catalog No. I09821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09821G 1 g
I0982200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09821G Supplier TCI America Supplier No. I09821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 288-16-4
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.124
MDL Number MFCD00020818
InChI Key ZLTPDFXIESTBQG-UHFFFAOYSA-N
Synonym isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone
PubChem CID 67515
ChEBI CHEBI:35600
IUPAC Name 1,2-thiazole
SMILES C1=CSN=C1

Specifications

Color Colorless
Boiling Point 114°C
Quantity 1 g
UN Number 1993
Formula Weight 85.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.