Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | ANSI Z87.1 ,CSA Z94.3-07 |
| Description | Dielectric |
| Certifications/Compliance (Europe) | EN-166 |
| Lens Tint | Indoor/Outdoor, Mirror |
| Frame Color | Black |
| Weight (Metric) | 41 g |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |