Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isovaleric acid, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018342 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8342 5 mL
11-101-9853 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8342 Supplier MilliporeSigma Supelco Supplier No. 786515ML
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 824°F
CAS 503-74-2
Vapor Pressure 0.38 mmHg (20°C)
Density 0.925 g/mL (at 20°C)
Flash Point 80°C - Pensky-Martens°Closed°Cup
Physical Form Neat
Linear Formula (CH3)2 CHCH2 COOH
MDL Number MFCD00002726
Melting Point -29°C
Boiling Point 175°C to 177°C
Synonym 3-Methylbutanoic acid; 3-Methylbutyric acid
Type Analytical Standard
Quantity 5 mL
Formula Weight 102.13
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.