Learn More
Cayman Chemical IsoverbascosIde 25mg
SDP
Supplier:  Cayman Chemical 2921625 MG
Encompass_Preferred
A phenylethanoid glycoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay and inhibits hydrogen peroxide-induced lipid peroxidation in V79-4 cells at 10 μg/ml; induces apoptosis and production of ROS in, and reduces viability of, OVCAR-3 cells from 7.5-30 μM; inhibits tumor growth in an OVCAR-3 mouse xenograft model at 30 mg/kg; decreases brain amyloid deposition and increases exploratory behavior in Aβ42-infused rats at 2.5 and 5 mg/kg; decreases xylene-induced EAr edema in mice and increases survival in a mouse model of LPS-induced endotoxic shock
Catalog No. 50-293-7194
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.