Learn More
Sigma Organic Chemistry isoxazole-4-boronic aci | 10mg | MFCD06657892
Supplier: Sigma Organic Chemistry CDS00704410MG

isoxazole-4-boronic aci, 10mg
About this Item:
MDL #: MFCD06657892
Molecular Weight: 112.88
UNSPSC Code: 12352200
Chemical Formula: C3H4BNO3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.