Learn More
Selleck Chemical LLC Ispinesib (SB-715992) 50mg 336113-53-2 CK0238273
SDP
Supplier:  Selleck Chemical LLC S145250MG
Encompass_Preferred
Ispinesib (SB-715992, CK0238273) is a potent, specific and reversible inhibitor of kinesin spindle protein (KSP) with Ki app of 1.7 nM in a cell-free assay, no inhibition to CENP-E, RabK6, MCAK, MKLP1, KHC or Kif1A. Ispinesib induces mitotic arrest and apoptotic cell death. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-857-8
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.