Learn More
3M™ IST-AIM Search and Rescue Kit
Catalog No. 19065628 Shop All 3M Products
Change view
Click to view available options
Search and Rescue Camera Kit
Catalog No.
19-065-628 Search and Rescue Camera Kit
1 options
Catalog No. 19-065-628 Supplier 3M™ Supplier No. 861210000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
  • Dotcam inspection camera
  • Portable full-color LCD monitor and attachable sunshield
  • Camera bracket with 8 ft. collapsible push-rod [collapses to 36 in. (91.4cm)]
  • 100 ft. (30m) cable
  • Impact- and water-resistant, foam-lined transport case.
Safely search inaccessible areas
Miniature Dotcam™ can access hazardous environments or areas too small for normal camera systems, robots, or rescuers. The camera can be secured onto an 8 ft. (2.4m) collapsible rod with a flexible gooseneck arm extension and sent deep into rubble to search for victims.
  • Rugged housing that's fully waterproof to 200 ft. depth
  • Compact 1 x 4 in. (2.5 x 10.2cm) high-resolution color camera with fixed focus and automatic gain
  • Six white LED lights as its own lighting source
Order Info
Replacement parts and optional accessories available separately. Contact your Customer Service Representative. Note: Dotcam not certified as intrinsically safe or explosion-proof.