Learn More
Apexbio Technology LLC ITE,10mg CAS# 448906-42-1
SDP
Supplier:  Apexbio Technology LLC A861510
Encompass_Preferred
2-(1′H-indole-3′-carbonyl)-thiazole-4-carboxylic acid methyl ester (ITE) is a nontoxic immunosuppressive endogenous aryl hydrocarbon receptor (AhR) ligand [1] [2]. Its EC50 identified via yeast AhR assay is 7.8×10−10 mol/l [3].AhR is a transcription factor mediating toxic effects of environmental pollutants such as 2, 3, 7, 8-tetrachlorodibenzo-p-dioxin (TCDD), it is activated by ligand [2]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0117
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.