Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITFG2 Monoclonal Antibody (OTI 3D8)
GREENER_CHOICE

Catalog No. PIMA525575
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA525575 100 μL
1 options

Catalog No. PIMA525575

Supplier: Invitrogen™ MA525575

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ITFG2 Monoclonal Antibody for Western Blot, Flow

As part of the KICSTOR complex functions in the amino acid-sensing branch of the TORC1 signaling pathway. Recruits, in an amino acid-independent manner, the GATOR1 complex to the lysosomal membranes and allows its interaction with GATOR2 and the RAG GTPases. Functions upstream of the RAG GTPases and is required to negatively regulate mTORC1 signaling in absence of amino acids. In absence of the KICSTOR complex mTORC1 is constitutively localized to the lysosome and activated. The KICSTOR complex is also probably involved in the regulation of mTORC1 by glucose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITFG2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI 3D8
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ITFG2
Gene Accession No. Q969R8
Gene Alias 2700050P07Rik; AI646725; integrin alpha FG-GAP repeat containing 2; integrin-alpha FG-GAP repeat-containing protein 2; integrin-alpha FG-GAP repeat-containing protein 2; KICSTOR complex protein ITFG2; ITFG2; KICSTOR complex protein ITFG2; MDS028; RGD1311944
Gene Symbols ITFG2
Host Species Mouse
Immunogen Full length human recombinant protein of ITFG2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55846
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.