Learn More
Invitrogen™ ITGA7 Monoclonal Antibody (334908), PE

Description
Reactivity with other Integrin alpha 7 isoforms was not tested. In direct ELISAs, no cross-reactivity with recombinant mouse Integrin alpha 2, alpha 3, alpha 4, alpha 5, alpha 9, alpha E, alpha X, recombinant human Integrin alpha L, or alpha 11 is observed.
Specifications
Specifications
| Antigen | ITGA7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 334908 |
| Conjugate | PE |
| Formulation | Buffered saline solution with 0.5% BSA and 0.09% sodium azide |
| Gene | Itga7 |
| Gene Accession No. | Q61738 |
| Gene Alias | [a]7; alpha 7A integrin; alpha7; H36-alpha7; integrin alpha 7; integrin alpha 7 chain; integrin alpha-7; Integrin alpha-7 70 kDa form; Integrin alpha-7 heavy chain; Integrin alpha-7 light chain; integrin subunit alpha 7; integrin, alpha 7; ITGA7; UNQ406/PRO768 |
| Gene Symbols | Itga7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.