Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549447 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549447 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549447 Supplier Invitrogen™ Supplier No. PA549447
  • $647.50
    $495.65 / Each of 1

    Save $151.85

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

ITGA7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Integrin alpha-7 is a protein that in humans is encoded by the ITGA7 gene. This gene is mapped to 12q13.2. The protein encoded by this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. They mediate a wide spectrum of cell-cell and cell-matrix interactions, and thus play a role in cell migration, morphologic development, differentiation, and metastasis. This protein functions as a receptor for the basement membrane protein laminin-1. It is mainly expressed in skeletal and cardiac muscles and may be involved in differentiation and migration processes during myogenesis. Defects in this gene are associated with congenital myopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA7
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Itga7
Gene Accession No. Q13683
Gene Alias [a]7; alpha 7A integrin; alpha7; H36-alpha7; integrin alpha 7; integrin alpha 7 chain; integrin alpha-7; Integrin alpha-7 70 kDa form; Integrin alpha-7 heavy chain; Integrin alpha-7 light chain; integrin subunit alpha 7; integrin, alpha 7; ITGA7; UNQ406/PRO768
Gene Symbols Itga7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 247-279 amino acids from the N-terminal region of human ITGA7.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3679
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.