Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547540
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547540 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547540 Supplier Invitrogen™ Supplier No. PA547540
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human Integrin alpha 1, alpha 8, alpha 9, alpha L, recombinant mouse Integrin 3, 4, 5, E, and M is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane glycoproteins composed of an alpha chain and a beta chain that mediate cell-cell and cell-matrix adhesion. The protein encoded by this gene, when bound to the beta 1 chain, forms an integrin that is a receptor for VCAM1, cytotactin and osteopontin. Expression of this gene has been found to be upregulated in small cell lung cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA9
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Itga9
Gene Accession No. B8JK39
Gene Alias (alpha)9; 2610002H11Rik; 6720458D17Rik; AI461869; ALPHA-RLC; D130073C02; D9Ertd428e; integrin alpha 9; integrin alpha-9; Integrin alpha-RLC; integrin subunit alpha 9; integrin, alpha 9; ITGA4L; Itga9; Itga9l; RLC
Gene Symbols Itga9
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Integin alpha 9 Try31-Val979.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104099
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.