Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB3BP Monoclonal Antibody (OTI4H1), TrueMAB™, OriGene
SDP

Catalog No. 501679758
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9758 Supplier OriGene Supplier No. TA803458
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ITGB3BP is a transcription coregulator that can have both coactivator and corepressor functions. Isoform 1, but not other isoforms, is involved in the coactivation of nuclear receptors for retinoid X (RXRs) and thyroid hormone (TRs) in a ligand-dependent fashion. ITGB3BP acts as a transcriptional corepressor via its interaction with the NFKB1 NF-kappa-B subunit, possibly by interfering with the transactivation domain of NFKB1. It induces apoptosis in breast cancer cells, but not in other cancer cells, via a caspase-2 mediated pathway that involves mitochondrial membrane permeabilization but does not require other caspases. ITGB3BP may also act as an inhibitor of cyclin A-associated kinase. ITGB3BP may be involved in incorporation of newly synthesized CENPA into centromeres via its interaction with the CENPA-NAC complex.

Specifications

Antigen ITGB3BP
Applications Western Blot
Classification Monoclonal
Clone OTI4H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ITGB3BP
Gene Accession No. Q13352
Gene Alias CENP-R, CENPR, HSU37139, NRIF3, TAP20
Gene Symbols ITGB3BP
Host Species Mouse
Immunogen Full length human recombit protein of human ITGB3BP produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23421
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.