Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB4 Rabbit anti-Human, Mouse, Rat, Polyclonal, liquid, OriGene
SDP

Catalog No. 501684816
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4816 Supplier OriGene Supplier No. TA890131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD104 (ITGB4) is an integrin alpha-6/beta-4 is a receptor for laminin. It plays a critical structural role in the hemidesmosome of epithelial cells. It is required for the regulations of keratinocyte polarity and motility. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Specifications

Antigen ITGB4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ITGB4
Gene Accession No. A2A863, P16144, Q64632
Gene Alias CD104, GP150
Gene Symbols ITGB4
Host Species Rabbit
Immunogen Recombit protein of human ITGB4
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192897, 25724, 3691
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.