Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547588
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547588 Supplier Invitrogen™ Supplier No. PA547588
Only null left
Add to Cart
Edge
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 35% cross-reactivity with recombinant mouse Integrin beta 6 is observed and less than 5% cross-reactivity with recombinant human (rh) Integrin beta 3 and rhIntegrin beta 5 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

ITGB6 (Integrin alpha-V/beta-6) is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalization of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGB6
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Itgb6
Gene Accession No. P18564, Q9Z0T9
Gene Alias 2210409C20Rik; 4831415H04Rik; AI1H; integrin beta 6; integrin beta-6; integrin subunit beta 6; integrin, beta 6; Itgb6
Gene Symbols Itgb6
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Integrin beta 6 Gly22-Asn707.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16420, 3694
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.