Learn More
Description
PA1-1755 detects HC1 from human and mouse samples. PA1-1755 has been successfully used in immunofluorescence and immunohistochemical staining of paraffin embedded tissue sections. The PA1-1755 immunogen is a synthetic peptide corresponding to residues Q V Q E S L P E L S N. This immunizing peptide (PEP-303) is available for neutralization and control experiments. PA1-1755 can be used with blocking peptide PEP-301.
Specifications
Specifications
| Antigen | ITIH1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | ITIH1 |
| Gene Accession No. | P19827, Q61702 |
| Gene Alias | CYCSP39; H1P; HCP39; IATIH; IGHEP1; inter-alpha (globulin) inhibitor, H1 polypeptide; inter-alpha trypsin inhibitor, heavy chain 1; inter-alpha-inhibitor heavy chain 1; inter-alpha-trypsin inhibitor complex component III; inter-alpha-trypsin inhibitor heavy chain 1; inter-alpha-trypsin inhibitor heavy chain H1; Intin1; ITI heavy chain H1; ITIH; Itih1; Itih-1; ITI-HC1; MGC126415; Serum-derived hyaluronan-associated protein; SHAP |
| Gene Symbols | ITIH1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
