Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITK Monoclonal Antibody (2F12), eBioscience™
GREENER_CHOICE

Catalog No. 14975282
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-975-282 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-975-282 Supplier Invitrogen™ Supplier No. 14975282
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody 2F12 recognizes human, mouse, and rat ITK. ITK (also known as Interleukin-2 inducible T-cell kinase, T-cell-specific kinase, Tyrosine-protein kinase Lyk, Emt, and Tsk) is a member of the Tec family of kinases that include Tec, Btk, Bmx, and Txk. Tec family kinases play an indispensable role in activation of phospholipase C and activation of calcium-dependent signaling cascades following antigen receptor triggering. ITK is highly expressed by T cells and is required for optimal TCR-induced PLCgamma1 activation. Mice congenitally lacking ITK show profound defects in T cell development and signaling. Defects in ITK are the cause of lymphoproliferative syndrome EBV-associated autosomal type 1 (LPSA1). ITK also plays a role in the development of various T cell mediated inflammatory disorders such as atopic dermatitis, psoriasis, and allergic asthma. Applications Reported: This 2F12 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy. Applications Tested: This 2F12 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using high pH antigen retrieval and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined...

ITK is a tyrosine kinase expressed in T-cells which contains both SH2 and SH3 domains. ITK is thought to play a role in T-cell proliferation and differentiation. The Tec family of non-receptor tyrosine kinases is composed of six proteins designated Tec, Itk (also known as Emt or Tsk), Btk (previously known as Atk, BPK or Emb), Bmx, Txk (also known as Rlk) and Dsrc28C. All members of the family contain SH3 and SH2 domains and, with the exception of Txk and Dsrc28C, also contain pleckstrin homology (PH) and Tec homology (TH) domains in their amino termini. Itk associates with CD28 and becomes activated subsequent to CD28 ligation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2F12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ITK
Gene Accession No. Q03526, Q08881
Gene Alias EMT; homolog of mouse T-cell itk/tsk; IL2 inducible T cell kinase; IL2 inducible T-cell kinase; IL-2-inducible T cell kinase; IL-2-inducible T-cell kinase; IL2-inducible T-cell kinase; interleukin-2-inducible T cell kinase; interleukin-2-inducible T-cell kinase; ITK; Kinase EMT; kinase TLK; LPFS1; LYK; MGC126257; MGC126258; PSCTK2; T-cell-specific kinase; Tcsk; Tlk; Tsk; Tyrosine-protein kinase ITK/TSK; tyrosine-protein kinase LYK
Gene Symbols ITK
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16428, 363577, 3702
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.