Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITM2B Monoclonal Antibody (OTI1C11)
GREENER_CHOICE

Catalog No. PIMA525513
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525513 Supplier Invitrogen™ Supplier No. MA525513
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ITM2B Monoclonal Antibody for Western Blot, IHC (P), Flow

Amyloid precursor proteins are processed by beta-secretase and gamma-secretase to produce beta-amyloid peptides which form the characteristic plaques of Alzheimer disease. This gene encodes a transmembrane protein which is processed at the C-terminus by furin or furin-like proteases to produce a small secreted peptide which inhibits the deposition of beta-amyloid. Mutations which result in extension of the C-terminal end of the encoded protein, thereby increasing the size of the secreted peptide, are associated with two neurogenerative diseases, familial British dementia and familial Danish dementia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITM2B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ITM2B
Gene Accession No. Q9Y287
Gene Alias ABRI; ABri/ADan amyloid peptide; ABri23; AI256040; Bri; BRI2; BRI2 ICD; BRI2 intracellular domain; BRI2, membrane form; Bri2-23; Bri23 peptide; BRI2C, soluble form; Bricd2b; BRICHOS domain containing 2B; C-terminal peptide; D14Sel6; E25B; E25BMM; E3-16; FBD; imBRI2; Immature BRI2; integral membrane protein 2B; Integral membrane protein 2B (E25B protein); ITM2B; Mature BRI2; mBRI2; OTTHUMP00000018394; P23 peptide; Protein E25B; RDGCA; RP11-196P14.1; Transmembrane protein BRI
Gene Symbols ITM2B
Host Species Mouse
Immunogen Full length human recombinant protein of ITM2B produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9445
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.