Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Itraconazole, 99%

Catalog No. AC452870050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452870050 5 g
AC452870250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452870050 Supplier Thermo Scientific Chemicals Supplier No. 452870050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Itraconazole, or voriconazole, CAS # 84625-61-6, is an N-arylpiperazine that has antifungal properties. | CAS: 84625-61-6 | C35H38Cl2N8O4 | 705.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Itraconazole is an aromatic ether that works as a strong inhibitor of P-glycoproteins and CYP3A4

• Itraconazole also acts as a hedgehog signaling inhibitor and P450 inhibitor

Applications

• Itraconazole can be exhibits antifungal activity against organisms including aspergillosis, blastomycosis, candidiasis, chromoblastomycosis, coccidioidomycosis, cryptococcosis, histoplasmosis, and sporotrichosis

• Itraconazole may be used to experimentally target the delivery of other molecules, such as plasmids, to cells

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84625-61-6
Molecular Formula C35H38Cl2N8O4
Molecular Weight (g/mol) 705.64
MDL Number MFCD00870168,MFCD00941396
InChI Key VHVPQPYKVGDNFY-ZPGVKDDISA-N
Synonym itraconazole-d5, itraconazole, oriconazole-d5, triasporin-d5, sporamelt-d5, itrizole-d5, sporanox-d5, +/--4-4-4-4-2r,4s-2,4-dichlorophenyl-2-1h-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl-1-piperazinyl phenyl-2,4-dihydro-2-1-methylpropyl-d5-3h-1,2,4-triazol-3-one, 2h5-4-4-4-4-2-1h-1,2,4-triazol-1-yl methyl-2-2,4-dichlorophenyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1-sec-butyl-1h-1,2,4-triazol-5 4h-one, 2-3,3,4,4,4-2 h? butan-2-yl-4-4-4-4-2r,4s-2-2,4-dichlorophenyl-2-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1,2,4-triazol-3-one
PubChem CID 45039617
IUPAC Name 4-[4-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-(3,3,4,4,4-pentadeuteriobutan-2-yl)-1,2,4-triazol-3-one
SMILES CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1

Specifications

Melting Point 168.0°C to 170.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 5 g
Merck Index 15,5292
Solubility Information Solubility in water: <1mg/mL. Other solubilities: solubility in ethanol: <1mg/mL, solubility in dmso: 4mg/mL
Formula Weight 705.64
Percent Purity 99%
Chemical Name or Material Itraconazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.