CAS RN 84625-61-6
Thermo Scientific Chemicals Itraconazole, 99%
CAS: 84625-61-6 Molecular Formula: C35H38Cl2N8O4 Molecular Weight (g/mol): 705.64 MDL Number: MFCD00870168,MFCD00941396 InChI Key: VHVPQPYKVGDNFY-ZPGVKDDISA-N Synonym: itraconazole-d5,itraconazole,oriconazole-d5,triasporin-d5,sporamelt-d5,itrizole-d5,sporanox-d5,+/--4-4-4-4-2r,4s-2,4-dichlorophenyl-2-1h-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl-1-piperazinyl phenyl-2,4-dihydro-2-1-methylpropyl-d5-3h-1,2,4-triazol-3-one,2h5-4-4-4-4-2-1h-1,2,4-triazol-1-yl methyl-2-2,4-dichlorophenyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1-sec-butyl-1h-1,2,4-triazol-5 4h-one,2-3,3,4,4,4-2 h? butan-2-yl-4-4-4-4-2r,4s-2-2,4-dichlorophenyl-2-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1,2,4-triazol-3-one PubChem CID: 45039617 IUPAC Name: 4-[4-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-(3,3,4,4,4-pentadeuteriobutan-2-yl)-1,2,4-triazol-3-one SMILES: CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1
Itraconazole, 98%
CAS: 84625-61-6 Molecular Formula: C35H38Cl2N8O4 Molecular Weight (g/mol): 705.64 MDL Number: MFCD00870168,MFCD00941396 InChI Key: VHVPQPYKVGDNFY-ZPGVKDDISA-N Synonym: Oriconazole; R51211 PubChem CID: 45039617 IUPAC Name: 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one SMILES: CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1
Itraconazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
MilliporeSigma™ Calbiochem™ Hh Signaling Antagonist X, Itraconazole
The Hh Signaling Antagonist X, Itraconazole, also referenced under CAS 84625-61-6, controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.
Itraconazole 98.0+%, TCI America™
CAS: 84625-61-6 Molecular Formula: C35H38Cl2N8O4 Molecular Weight (g/mol): 705.64 MDL Number: MFCD00870168,MFCD00941396 InChI Key: VHVPQPYKVGDNFY-ZPGVKDDISA-N Synonym: itraconazole-d5,itraconazole,oriconazole-d5,triasporin-d5,sporamelt-d5,itrizole-d5,sporanox-d5,+/--4-4-4-4-2r,4s-2,4-dichlorophenyl-2-1h-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl-1-piperazinyl phenyl-2,4-dihydro-2-1-methylpropyl-d5-3h-1,2,4-triazol-3-one,2h5-4-4-4-4-2-1h-1,2,4-triazol-1-yl methyl-2-2,4-dichlorophenyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1-sec-butyl-1h-1,2,4-triazol-5 4h-one,2-3,3,4,4,4-2 h? butan-2-yl-4-4-4-4-2r,4s-2-2,4-dichlorophenyl-2-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1,2,4-triazol-3-one PubChem CID: 45039617 IUPAC Name: 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one SMILES: CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1
R&D Systems™ Itraconazole
A Tocris product, part of R&D Systems. SMO antagonist; acts at different binding site to cyclopamine (Cat No. 1623)
Itraconazole, Micronized, USP, 98.5-101.5%, Spectrum™ Chemical
CAS: 84625-61-6 Molecular Formula: C35H38Cl2N8O4 Molecular Weight (g/mol): 705.64 MDL Number: MFCD00870168,MFCD00941396 InChI Key: VHVPQPYKVGDNFY-ZPGVKDDISA-N IUPAC Name: 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one SMILES: CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1