Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Itraconazole, Cytochrome P450 Inhibitor, >99% Purity, Solid, 100 mg
Description
- Molecular Weight: 705.64 g/mol
- Purity: >99%
- Form: Solid
- Activity: Cytochrome p450 CYP3A4 inhibitor (IC50 = 16-26 nM for midazolam hydroxylation)
- Selectivity: Higher affinity for fungal enzymes over mammalian cytochrome p450
- Properties: Limited ability to cross the blood-brain barrier; antifungal activity
- Composition: 100% Itraconazole
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Cytochrome P450 (CYP3A4) |
| Shipping Condition | Ambient |
| Buffer | 100% Itraconazole |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% Itraconazole |
| Quantity | 100 mg |
| For Use With (Application) | Cytochrome p450 inhibition, Antifungal activity |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?