Learn More
Abcam Itraconazole, Cytochrome P450 Inhibitor, >99% Purity, Solid, 50 mg
Description
- Molecular weight: 705.64 g/mol
- Purity: >99%
- Form: Solid
- Activity: Cytochrome p450 CYP3A4 inhibitor (IC50 = 16 to 26 nM for midazolam hydroxylation)
- Selectivity: Higher affinity for fungal enzymes over mammalian cytochrome p450
- Biological properties: Limited ability to cross the blood-brain barrier; Antifungal activity
- Composition: 100% Itraconazole
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Cytochrome p450 (CYP3A4) |
| Buffer | 100% Itraconazole |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% Itraconazole |
| Quantity | 50 mg |
| For Use With (Application) | Cytochrome p450 inhibition, Antifungal activity |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.