Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itraconazole, Micronized, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603593 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-593 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-603-593 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I26621KGBL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

I2662, 84625-61-6, C35H38Cl2N8O4

Spectrum™ Chemical Itraconazole, Micronized, USP is a drug used for the treatment of fungal infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 84625-61-6
Molecular Formula C35H38Cl2N8O4
Molecular Weight (g/mol) 705.64
MDL Number MFCD00870168,MFCD00941396
InChI Key VHVPQPYKVGDNFY-ZPGVKDDISA-N
IUPAC Name 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one
SMILES CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 1 kg
Optical Rotation -0.10° to +0.10°
Percent Purity 98.5 to 101.5%
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Itraconazole, Micronized,
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.