Learn More
Sigma Aldrich Fine Chemicals Biosciences iTRAQR Reagent 8PLEX Mul
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 43816631KT
Encompass
Contains iTRAQ(R) Reagents 113 114 115 116 117 118 119 and 121 and sufficient material for five 8-plex assays. Multiplex Buffer kit (Cat. No. 4381664) to be ordered separately.Cat. No. 4381663 contains the equivalent reagents in the Sciex iChemistry(R) product number 4390812.
Catalog No. 50-290-8947
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.