Learn More
R&D Systems™ ITS Media Supplement (100X)
GSA_VASDP
Catalog No. AR013 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
AR013 5 mL
1 options
This item is not returnable. View return policy
Catalog No. AR013 Supplier R&D Systems™ Supplier No. AR013
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Supports cell proliferation under low serum conditions. R&D Systems ITS Media Supplement (100X) is a pre-optimized and serum-free media supplement that helps to maintain culture media consistency across passages and experiments.
  • Contains optimized concentrations of insulin, transferrin and selenite
  • Chemically-defined media decreases experimental variation
  • High lot-to-lot consistency decreases variation
  • Performance tested on multiple cell lines
  • QC-tested on multiple cell lines
Precautions
  • Transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen
  • As no testing can offer complete assurance of freedom from infectious reagents, these reagents should be handled as if capable of transmitting infection
Limitations
  • The safety and efficacy of the product in diagnostic or other clinical uses has not been established
  • The reagent should not be used beyond the expiration date indicated on the label
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Content And Storage 5mL ITS Media Supplement (100X). Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Type ITS (Insulin, Transferrin, Selenium)
Product Line R&D Systems
Quantity 5 mL
Regulatory Status RUO
For laboratory research use only. Not for use in diagnostic procedures.