Learn More
Med Vet International Terumo Surflo IV Catheter, 16G x 1.25in
Supplier:  Med Vet International SROX1632CA
Encompass_Preferred
The Terumo Surflo is a 1.25in peripheral IV equipped with an ultra-sharp double-bevel 16-gauge introducer needle coated with a medical-grade lubricant to help prevent inflammation as much as possible. Used to administer medicine intravenously. This is a flexible catheter with a large inner-wall diameter assuring good, stable blood flow. A preview chamber in clear sure-grip gives immediate indication of proper placement. Low risk of split tips and peel back. Comes individually in easy to handle, tamper-evident packaging.
Catalog No. 50-209-2357
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.