Learn More
Health Care Logistics Inc IV COVERS, LIGHT AMBER, 8 X 14
SDP
Supplier:  Health Care Logistics Inc 7589
Encompass
UV Blocking - Covers block 90% of UV light and are perfect for dispensing IV admixtures that contain light-sensitive drugs.Opacity - IV Covers are Light Amber with slit at sealed endTransparent - Bags are transparent, but will mask many differences in IV fluid color.Thickness - 1.5-mil - 1.8 milQuantity - 100 per packDimensions: 8"W x 14"H
Catalog No. NC1933975
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.