Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ivabradine hydrochloride
GREENER_CHOICE

Catalog No. 654210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-421-0 10 mg
65-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-421-0 Supplier Tocris Bioscience™ Supplier No. 6542/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

HCN channel blocker; inhibits If pacemaker current

Ivabradine hydrochloride is a HCN channel blocker (IC50 is approximately 0.5 - 2.5 μM). Bradycardic agent that inhibits If pacemaker current in sinoatrial node cells.

Specifications

Chemical Name or Material 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-2H-3-benzazepin-2-one hydrochloride
CAS 148849-67-6
Quantity 10 mg
Molecular Formula C27H36N2O5.HCl
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.