Learn More
Selleck Chemical LLC Ivacaftor (VX-770) 5mg 873054-44-5
SDP
Supplier:  Selleck Chemical LLC S11445MG
Encompass_Preferred
Ivacaftor (VX-770) is a selective potentiator of CFTR targeting G551D-CFTR and F508del-CFTR with EC50 of 100 nM and 25 nM in fisher rat thyroid cells, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0375409
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.