Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IVD Monoclonal Antibody (04)
GREENER_CHOICE

Catalog No. PIMA529374 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529374 Supplier Invitrogen™ Supplier No. MA529374
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Isovaleryl-CoA dehydrogenase/IVD.

Isovaleryl-CoA dehydrogenase (IVD) is a mitochondrial matrix enzyme that catalyzes the third step in leucine catabolism. The genetic deficiency of IVD results in an accumulation of isovaleric acid, which is toxic to the central nervous system and leads to isovaleric acidemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IVD
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IVD
Gene Accession No. P26440
Gene Alias 1300016K07Rik; 6720455E18Rik; ACAD2; AI463340; butyryl-CoA dehydrogenase; EGK_17362; isovaleryl CoA dehydrogenase; isovaleryl coenzyme A dehydrogenase; isovaleryl dehydrogenase; isovaleryl-CoA dehydrogenase; isovaleryl-CoA dehydrogenase, mitochondrial; IVD
Gene Symbols IVD
Host Species Mouse
Immunogen Recombinant Human Isovaleryl-CoA dehydrogenase/IVD protein (His30-His423).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3712
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.