Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Ivermectin, α7 Nicotinic Acetylcholine Receptor Modulator, Solid, 250 mg
Description
- Molecular Weight: 875.1 Da
- Purity: >96%
- Physical Form: Solid
- Activity: Positive allosteric modulator of α7 neuronal nicotinic acetylcholine receptors and P2X4 receptors; antihelmintic; modulates glutamate- and GABA-activated chloride channels; potentiates glycine-gated currents at low concentrations (30 nM)
- Storage Buffer: 100% Ivermectin
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Buffer | 100% Ivermectin |
| Shipping Condition | Ambient |
| Target Molecule | Alpha7 Nicotinic Acetylcholine Receptor, P2X4 Receptor, Glutamate-activated Chloride Channels, GABA-activated Chloride Channels, Glycine Receptor |
| Formulation | 100% Ivermectin |
| Quantity | 250 mg |
| For Use With (Application) | Neuroscience-alpha7 nicotinic acetylcholine receptors, Neuroscience-P2X4 receptors, Antihelmintic research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?