Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ivermectin, Tocris Bioscience™
GSA_VASDP

Catalog No. 1260100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-601-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-601-00 Supplier R&D Systems Supplier No. 1260/100

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Allosteric modulator of α7 nicotinic receptors

Positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor and the purinergic P2X4 receptor. Antihelmintic and antiviral. Also modulates glutamate- and GABA-activated chloride channels. Potentiates glycine-gated currents at low concentrations (30 nM). Inhibits replication of SARS-CoV-2 in vitro (IC50 ∽ 2μM).

This product is composed of both Ivermectin B1a (>90%) and Ivermectin B1b.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 70288-86-7
Molecular Formula C48H74O14
Molecular Weight (g/mol) 875.106
InChI Key AZSNMRSAGSSBNP-RDKIRRGNSA-N
Synonym ivermectin
PubChem CID 122172888
SMILES CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C

Specifications

Quantity 100 mg
Formula Weight 875.1
Percent Purity >96%
Chemical Name or Material Ivermectin
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.