Learn More
Cytek Biosciences Inc IVR MS CD28 (37.51) 500UG
Supplier:  Cytek Biosciences Inc 400281U500
Encompass_Preferred
In Vivo Ready Anti-Mouse CD28 (37.51) Isotype: Golden Syrian Hamster IgG Apps: FC, FA, IP Size: 500 ug
Catalog No. 50-105-4569
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.