Learn More
STEMCELL Technologies IWP-4, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72554
Encompass_Preferred
Inhibitor of WNT Production-4 (IWP-4) inhibits WNT signaling by inactivating Porcupine, a membrane-bound O-acyltransferase responsible for palmitoylating WNT proteins, which is essential for WNT signaling ability and secretion (Chen et al.). IWP-4 impairs WNT pathway activity in vitro with an IC₅₀ value of 25 nM (Chen et al.).DIFFERENTIATION· Promotes cardiomyocyte differentiation in human pluripotent stem cells after treatment with CHIR99021 (Lian et al., 2012, 2013; Sequiera et al.).· Promotes cardiomyocyte differentiation in human embryonic stem cells following primitive streak induction with BMP4 and Activin A (Hudson et al.).
Catalog No. 50-197-5942
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.