Learn More
TARGETMOL CHEMICALS INC IXA4 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T9349
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IXA4 is a highly selective nontoxic activator of IRE1/XBP1s and reduces APP secretion by activating IRE1. IXA4-stimulated IRE1 activation also enhanced pancreatic function. purity: 98%
Catalog No. 50-257-6205
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.