Learn More
Selleck Chemical LLC Ixabepilone (BMS-247550) 5mg 219989-84-1 Azaepothilone B, BMS 247550, BMS 247550-1, Ixempra
SDP
Supplier:  Selleck Chemical LLC S79305MG
Encompass_Preferred
Ixabepilone (BMS-247550, Azaepothilone B, BMS 247550-1, Ixempra) is an orally bioavailable microtubule inhibitor. It binds to tubulin and promotes tubulin polymerization and microtubule stabilization, thereby arresting cells in the G2-M phase of the cell cycle and inducing tumor cell apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5431
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.