Learn More
TARGETMOL CHEMICALS INC SW033291 10MG
Supplier: TARGETMOL CHEMICALS INC T2121
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SW033291 is a small-molecule inhibitor of 15-PGDH (Ki=0.1 nM) that increases prostaglandin PGE2 levels in bone marrow and other tissues. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.