Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JAB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589216 Supplier Invitrogen™ Supplier No. PA589216
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAASGSGMAQ KTWELANNMQ EAQSIDEIYK YDKKQQQEIL AAKPWTKDHH YFKYCKISAL ALLKMVMHAR SGGNLEVMGL MLGKVDGETM IIMDSFALPV EGTETRVNAQ AAAYEYMAAY IENAKQVGRL ENAIGWYHSH PGYGCWLSGI DVSTQMLNQQ FQEPFVAVVI DPTRTISAGK VNLGAFRTYP KGYKPPDEGP SEYQTIPLNK IEDFGVHCKQ YYALEVSYFK SSLDRKLLEL LWNKYWVNTL SSSSLLTNAD YTTGQVFDLS EKLEQSEAQL GRGSFMLGLE THDRKSEDKL AKATRDSCKT TIEAIHGLMS QVIKDKLFNQ INIS; Positive Samples: NIH/3T3, MCF-7, PC-12, HeLa, K-562, DU145, Mouse brain, Mouse heart, Mouse testis; Cellular Location: Cytoplasm, Cytoplasmic vesicle, Nucleus, perinuclear region, secretory vesicle, synaptic vesicle.

JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the Signalosome, which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAB1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cops5
Gene Accession No. O35864, Q92905
Gene Alias 38 kDa Mov34 homolog; AI303502; COP9 (constitutive photomorphogenic) homolog, subunit 5; COP9 (constitutive photomorphogenic) homolog, subunit 5 (Arabidopsis thaliana); COP9 complex S5; COP9 constitutive photomorphogenic homolog subunit 5; COP9 signalosome complex subunit 5; COP9 signalosome subunit 5; Cops5; CSN5; JAB1; Jun activation domain-binding protein 1; Jun coactivator; Kic2; Kip1 C-terminus-interacting protein 2; MGC3149; Mov34; MOV-34; SGN5; signalosome subunit 5; testis secretory sperm-binding protein Li 231m
Gene Symbols Cops5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-334 of human JAB1/CSN5/JAB1/CSN5/COPS5 (NP_0068282).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10987, 26754, 312916
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.