Learn More
Strategic Applications Inc Large Animal Jacket Pocket for SAI Mini Ambulatory Infusion Pump
Supplier: Strategic Applications Inc JP MINI
This item has a minimum qty of 5 per supplier requirements.
The JP MINI jacket pocket is perfectly sized for use with the SAI 3D Mini Ambulatory Pump at 7" x 2.75" x 2". Inquire with SAI to have the pocket attached to a jacket. Pockets are ideal for holding ambulatory pumps, medication reservoirs, administration sets, or other apparatus that must travel with the animal in preclinical research studies. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply. Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.