Learn More
Description
- Round bottom flask with standard taper outer joints on the center and vertical side necks
- Heavy wall design
- Entire flask body is jacketed and has two hose connections for circulation of heating or cooling liquid.
Specifications
Specifications
| Color | Clear |
| Capacity (Metric) | 1 L |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Organic Synthesis |
| Standard Taper Joint | Center: 29/42; Side: 24/40 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

