Learn More
Jaece Industries Identi-plug™ Plastic Foam Stoppers
For test tubes and flasks
$117.70 - $274.50
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
14-127-40D
|
Jaece Industries
L800C |
3.4 cm | 4.44 cm |
Case of 200 for $182.50
|
|
||||
14-127-40A
|
Jaece Industries
L800A |
1.3 cm | 3.5 cm |
Case of 1000 for $204.00
|
|
||||
14-127-40E
|
Jaece Industries
L800D |
4.5 cm | 4.44 cm |
Case of 100 for $132.80
|
|
||||
14-127-40F
|
Jaece Industries
L800D2 |
5 cm | 5.63 cm |
Case of 80 for $184.60
|
|
||||
14-127-40B
|
Jaece Industries
L800B |
2 cm | 3.5 cm |
Case of 1000 for $274.50
|
![]() |
||||
14-127-40G
|
Jaece Industries
L800E |
6.5 cm | 4.44 cm |
Case of 50 for $117.70
|
|
||||
14-127-41
|
Jaece Industries
L800G |
7.5 cm | 4.44 cm |
Case of 40 for $136.00
|
|
||||
14-127-42
|
Jaece Industries
L800E2 |
6.5 cm | 8.89 cm |
Case of 30 for $121.00
|
|
||||
Description
For test tubes and flasks
- Easily compressed to form a snug-fitting stopper
- When removed from container, they spring back to original shape
- Permit air exchange and can be used over a wide range of temperatures; ideal for use with drosophilia vials
- Withstand constant use and sterilization
- Autoclavable and chemically resistant
Specifications
Autoclavable | |
White | |
Plug Stopper |