Learn More
AnaSpec JAG-1 (188-204) NOTCH LIGAND
Supplier:  AnaSpec AS61298
Encompass
JAG-1 (188-204), Jagged-1 (188-204), Notch Ligand, DSL Peptide[CDDYYYGFGCNKFCRPR] ; MW 2107.4; (1 mg) This peptide is a fragment of the JAG-1 protein. JAG-1 is Notch ligand, a peptide that is the most conspicuously expressed ligand in skin. JAG-1 induces epidermal maturation. Exposing submerged keratinocytes monolayers to JAG-1 with elevated calcium concentration produces stratification with loricrin expression and NF-κB activation.
Catalog No. NC0243900
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.