Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Jagged 2 Monoclonal Antibody (HMJ2-1), eFluor™ 660, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7218871
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-339-282 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-339-282 Supplier Invitrogen™ Supplier No. 50339282
Only null left
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

This HMJ2-1 MAb reacts with mouse Jagged-2, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g., Notch 1-4), Jagged-2 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Jagged-2 is expressed by thymic lymphoid and stromal cells, as well as macrophages and dendritic cells in the spleen. Note: Staining of endogenous splenic dendritic (CD8+CD11c+) cells could only be consistently confirmed in cells isolated from C57BL/6, but not Balb/c, mice. Other strains have not been tested. The antiobdy has been reported to crossreact to rat and have blocking function.

The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. JAG2 is one of several ligands that activate Notch and related receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Jagged 2
Applications Flow Cytometry
Classification Monoclonal
Clone HMJ2-1
Concentration 0.2 mg/mL
Conjugate eFluor 660
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Jag2
Gene Accession No. P97607, Q9QYE5
Gene Alias D12Ggc2e; HJ2; JAG2; jagged 2; Jagged2; mJagged2-1; protein jagged-2; SER2; Serh; sm; syndactylism
Gene Symbols Jag2
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16450, 29147
Target Species Mouse, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.