Learn More
Epigentek Inc JAK3 Polyclonal Antibody, Biotin Conjulated (100 ul)
SDP
Supplier:  Epigentek Inc A59499100
Encompass_Preferred
FITC Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Tyrosine-protein kinase JAK3 protein (351-614AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Tyrosine-protein kinase JAK3 (EC:2.7.10.2), JAK3, Janus kinase 3, JAK-3, Leukocyte janus kinase, L-JAK.
Catalog No. 50-123-3080
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.