Learn More
Invitrogen™ JAM3 (soluble) Monoclonal Antibody (19H36)

Description
MA1-40059 detects JAM-C from mouse and human samples. MA1-40059 has been successfully used in flow cytometry, immunohistochemistry (frozen), ELISA and immunoprecipitation applications. The MA1-40059 immunogen is mouse JAM-C.
Specifications
Specifications
| Antigen | JAM3 (soluble) |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 19H36 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | JAM3 |
| Gene Accession No. | Q9BX67, Q9D8B7 |
| Gene Alias | 1110002N23Rik; JAM-2; JAM3; JAM-3; JAMC; JAM-C; Jcam3; junction adhesion molecule 3; junction cell adhesion molecule 3; junction cell adhesion molecule C; junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C; UNQ859/PRO1868 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.