Learn More
Description
Key information: Janelia Fluor® 585, Maleimide is an orange fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity). Suitable for live cell imaging.
Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells) and STED. Cell permeable.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 585 nm and 609 nm, respectively; quantum yield = 0.78; extinction coefficient = 156,000 M-1cm-1(measured in Ethanol or TFE plus 0.1% TFA); A280 correction factor = 0.099; lactone-zwitterion equilibrium constant (KL-Z) < 0.0001.
Please see the protocol for further information and a guide to protein/antibody labeling.
Janelia Fluor® is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Quantity | 1 mg |
| Content And Storage | Store at -20°C |
| Target | Sulfhydyl-Reactive Janelia Fluor Dyes |
| Product Line | Tocris |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
